You searched for: Homogenizers
A homogenizer is used for the proper mixing of non-soluble liquids to have the same throughout. Commonly used for sample preparation, the laboratory equipment pieces allow nucleic acids, proteins, cells, metabolism, pathogens, and other target analyses to be accurate. Although different methods may be used, the modern devices guarantee that the material consistency is uniform. Fast-acting homogenizers have sophisticated microprocessors to tailor cell disrupting operations to each specific sample being handled.
VWR® 250 Homogenizer
Supplier: VWR International
The VWR® 250 Homogenizer unit is a perfect fit between the VWR® 200 Homogenizer unit and the benchtop VWR® Homogenizer units.
Expand 1 Items
VWR® Bead Mill Homogenizer, 120 V
Supplier: VWR International
The VWR® Bead Mill is part of the newest generation of Bead Mill homogenizers. This economical system is specifically designed for grinding, lysing and homogenization of up to 16 samples. Using sample tubes pre-filled with a variety of lysing beads, the VWR® Bead Mill vigorously and uniformly shakes the tubes providing an efficient, consistent, high-yield and quality homogenization usually in less than 40 seconds.
Expand 1 Items
VWR® 25D Digital Homogenizer
Supplier: VWR International
VWR® 25D Digital Homogenizer
Expand 1 Items
VWR® Micro Homogenizing Packages
Supplier: VWR International
The VWR® Standard Micro-Homogenizing Package fits into 500µL, 1.5 mL, and 2 mL micro-tubes.
Expand 3 Items
VWR® Premium GenPack Homogenizing Package
Supplier: VWR International
The VWR® Premium GenPack Homogenizing Package is designed for homogenizing multiple samples quickly and easily.
Expand 1 Items
VWR® 400D Digital Benchtop Homogenizer
Supplier: VWR International
The VWR® 400D Digital Benchtop Homogenizer unit is the optimal balance of power and superior performance, combining a high-watt motor and advanced technology in digital speed controls to result in the most reliable and durable laboratory benchtop homogenizer unit.
Expand 1 Items
VWR® Ultrasonic Homogenizers
Supplier: VWR International
VWR® Ultrasonic Homogenizers are next generation sonicators designed to simplify the process of particle size reduction and cell lysis. Designed for research labs that require stringent parameter control, reproducibility, and programmability, the VWR® Ultrasonic Homogenizers feature an intuitive touch screen interface.
Expand 2 Items
VWR® 4-Place Mini Bead Mill Homogenizer
Supplier: VWR International
The VWR® 4-Place Mini Bead Mill Homogenizer is a compact and powerful bead mill homogenizer.
Expand 1 Items
VWR® 400DEL Digital Benchtop Homogenizer
Supplier: VWR International
The VWR® 400DEL Digital Benchtop Homogenizer unit is the optimal balance of power and superior performance, combining a high-watt motor and advanced technology in digital speed controls to result in the most reliable and durable laboratory benchtop homogenizer unit.
Expand 1 Items
VWR® MaX Homogenizing Packages
Supplier: VWR International
The VWR® Standard MaX-Homogenizing Package is ideal for homogenizing in various tube sizes, from 10 mL to 50 mL.
Expand 3 Items
VWR® Universal Homogenizing Packages
Supplier: VWR International
The VWR® Standard Universal Homogenizing Package is designed for use with all types of samples, even tough or stringy.
Expand 2 Items
VWR® Disposable Pellet Mixers and Cordless Motor
Supplier: VWR International
Designed to homogenize cells and plant tissue or to re-suspend protein and DNA pellets.
Expand 7 Items
Tissue Homogenizer, Dounce, Ace Glass Incorporated
Supplier: Ace Glass
This Dounce tissue homogenizer is supplied with two pestles, one "loose" and one "tight", to ensure dissociation of cells into fine particles with minimal damage to cell nuclei
Expand 4 Items
Tenbroeck Tissue Grinders, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
For homogenizing delicate tissues such as intestines, liver, and heart
Expand 5 Items
Potter-Elvehjem Tissue Grinders, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Grinders for preparation of soft tissue homogenates, such as liver, brain.
Expand 19 Items
Tissue Homogenizer, Ace Glass Incorporated
Supplier: Ace Glass
This highly efficient, all glass, tapered tissue grinder/homogenizer combines two grinding surfaces on a single pestle and tube
Expand 12 Items
Safe-Grind® Plastic-Coated Tissue Grinders with Fluoropolymer Resin Pestles, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
For sure grip and safe grinding of homogenate even with power homogenization.
Expand 5 Items
Tissue Homogenizer, Potter-Elehjem, Ace Glass Incorporated
Supplier: Ace Glass
When using this Potter-Elehjem tissue homogenizer, homogenization occurs as the sample and buffer are forced through the cylindrical portion of the mortar as the pestle is rotated downward
Expand 6 Items
KIMBLE® DUALL® Tissue Grinders, PTFE Pestles and Glass Tubes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Tissue grinders greatly improve grinding efficiency by combining both conical and cylindrical surfaces to effectively reduce tissue and produce a uniform homogenate
Expand 4 Items
KONTES® Potter-Elvehjem Tissue Grinders with PTFE Pestles, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
For homogenization of tissue samples
Expand 7 Items
Tissue Homogenizer, Potter-Elvehjem, Micro, Ace Glass Incorporated
Supplier: Ace Glass
This micro size tissue homogenizer is for extremely precise work
Expand 1 Items
Micro Tissue Grinder, Disposable, Wheaton
Supplier: DWK Life Sciences
The Disposable Micro Tissue Grinder allows for homogenized tissue micro samples without cross-contamination
Expand 1 Items
PYREX® Potter-Elvehjem Tissue Grinder with Fluoropolymer Resin Pestles, Corning
Supplier: Corning
Homogenizers designed for preparing small samples for pathological and biochemical studies.
Expand 7 Items
Tapered Tissue Grinders, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Grinders are available with glass or fluoropolymer resin pestles
Expand 7 Items
Micro-Tissue Grinder Kit, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Micro tissue grinder kit is ideal for sterile homogenization processes.
Expand 1 Items
Dounce Tissue Grinders, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Designed for fine particle size reduction without damage to cell nuclei
Expand 4 Items
Extraction Kits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Extraction kits with optional ceramic homogenizers feature salts for buffering of samples.
Expand 3 Items
KIMBLE® DUALL® Tissue Grinders, All-Glass with Screw-Cap, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
The DUALL® is an extremely efficient grinder which combines both conical and cylindrical surfaces on a single pestle and tube. These separate areas perform the dual functions of initial grinding in the conical section and final homogenization in the cylindrical section.
Expand 3 Items
Kimble® Potter-Elvehjem Tissue Grinder Components, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
A graduated glass tube and a finely machined PTFE pestle comprise this popular style.
Expand 8 Items
Tissue Homogenizer, Tenbroeck, Ace Glass Incorporated
Supplier: Ace Glass
These all-glass Tenbroeck tissue grinders come with precision-made, interchangeable pestles and tubes