ContactUsLinkComponent
PD MultiTrap™ G-25, Filter Plates, 96-Well, Cytiva
PD MultiTrap™ G-25, Filter Plates, 96-Well, Cytiva
Catalog # CA95056-248L
Supplier:  Cytiva
undefined
PD MultiTrap™ G-25, Filter Plates, 96-Well, Cytiva
Catalog # CA95056-248L
Supplier:  Cytiva
Supplier Number:  28-9180-06
Specifications
  • No. of wells:
    96
  • Description:
    PD MultiTrap™ G-25
  • Cat. no.:
    CA95056-248L
  • Supplier No.:
    28-9180-06
Specifications
About this item
PD MultiTrap G-25 is designed for high-throughput desalting and buffer exchange of multiple samples.
  • Convenient and fast small-scale cleanup of proteins and other large biomolecules (>5000 Mr) with high reproducibility well-to-well and plate-to-plate
  • Range of applications such as desalting, buffer exchange, and removal of low-molecular weight compounds
  • High desalting capacity
  • Suitable for manual use as well as for automation together with robotic systems
  • Sample preparation in the range of 70 to 130 μl without sample dilution
PD MultiTrap G-25 is prepacked with 500 µl Sephadex G-25 resin/well for high reproducibility. The 96-well plates are manufactured frombiocompatible polypropylene.