SOURCE™ 15RPC Reversed Phase Chromatography Media, Cytiva
Catalog # CA89128-850
Supplier: Cytiva
CAS Number:
Specifications
- Particle Size:~15 µm
- Description:SOURCE™ 15RPC
- Size:10 ml
- pH range:1 - 12
- Matrix:Polystyrene/divinylbenzene
- Ligand:Matrix surface
- Storage temperature:4...30 °C
- Binding capacity:~10 mg BSA/ml at 300 cm/h, ~30 mg bacitracin/ml at 300 cm/h, ~50 mg insulin/ml at 300 cm/h
- Cat. no.:CA89128-850
- Type:Liquid
- Supplier No.:17-0727-20
Specifications
About this item
SOURCE™ 15RPC is designed for reversed phase chromatography (RPC) with full pH range stability and suitable for polishing steps in industrial processes requiring high flow velocities and low back-pressure.
- Mono-sized 15 µm rigid beads gives low back pressures and high flow rates allowing for high productivity
- Designed for high resolution reversed phase chromatography of proteins, peptides and oligonucleotides
- High chemical stability allowing for wide range of working conditions with good resistance to cleaning conditions at high pH
- Bioprocess medium supported for industrial applications
A controlled pore size distribution and large specific surface area offer excellent capacity for a wide range of molecules, from small peptides and oligonucleotides up to large proteins. With full pH range stability, SOURCE™ 15RPC is a strong alternative to silica-based RPC matrices.
Available in a range of different bulk pack sizes for easy scale-up and process development.