Specifications
- Method / Format:PrecipitationColumn (silica-based)
- Prep size:Mini
- Refine target molecule:Total RNATotal DNATotal protein
- Sample type:Cultured cellsHuman/animal cells and tissues
- Target molecule:DNARNAProtein
- Description:illustra™ triplePrep Kit
- Size:50 Preps
- Description:illustra™ triplePrep Kit
- Cat. no.:CA95056-354L
- Supplier No.:28-9425-44
Specifications
About this item
The illustra™ triplePrep Kit is designed for the rapid, simultaneous extraction and isolation of high yield genomic DNA (gDNA), total RNA, and total denatured proteins from undivided animal tissues and mammalian cells. The streamlined workflow reduces the overall number of steps, enabling the preparation of all three analytes in less than one hour.
- High-quality genomic DNA with a greater proportion of higher molecular weight DNA and less shearing
- Demonstrated application to various types of whole blood including human, horse, rabbit, rat, and mouse
- Increased ease of use with reduced pipetting volume changes, one centrifugation setting, and color-coded kit components
- Verified in several downstream applications including real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests
The purification process involves minimal shearing resulting in the production of 5 to 10 μg of good quality, intact genomic DNA from a 200 μl sample. Input sample volumes can vary from 50 μl to 1 ml. The method uses a simple genomic DNA purification protocol that uses chaotropic agents to extract DNA from blood cells, denature protein components, and promote the selective binding of DNA to a column-based, novel silica membrane. The elution volume can be modified as necessary to obtain the appropriate genomic DNA concentration for most downstream molecular biological applications.