You searched for: Cell Culture Flasks
VWR supplies cell culture flasks in a variety of sizes for optimal cultivation of cells in your lab. Select a polystyrene flask with a canted, angled, or straight neck to suit your purpose. Products feature vented, plug-seal, or phenolic style caps for tight closure. Sterile and cleanroom-assembled flasks with printed volume gradations or raised rims for stacking are offered. Cell growth areas and flask volumes are available in an array of sizes. Flasks may be ordered in packs or cases.
VWR® Cell Culture Flasks Treated for Increased Cell Attachment, Sterile
Supplier: VWR International
Increased Cell Attachment surface treatment can improve cell spreading and attachment
Expand 10 Items
VWR® Cell Culture Flasks
Supplier: VWR International
VWR® Cell culture flasks are vacuum gas plasma treated for consistent cell attachment and growth.
Expand 10 Items
VWR® Tissue Culture Flask, Non-Treated, Sterilized, Non-Pyrogenic
Supplier: VWR International
Cell and Tissue Culture Flasks are ideal for cell growth and cell yields.
Expand 12 Items
Avantor Sterile Polycarbonate Single-Use Flask Assemblies, Avantor Fluid Handling
Supplier: VWR International
Sterile polycarbonate flasks with weldable tubing provide the maximum amount of flexibility and reliability.
Expand 5 Items
Corning® Polystyrene Tissue Culture Flasks, Sterile, Corning
Supplier: Corning
Designed and treated for optimal cell attachment and growth
Expand 7 Items
Corning® Cell Culture Flasks with Vent Cap, Non-Treated, Corning
Supplier: Corning
These culture flasks are constructed from optically clear virgin polystrene and feature a canted neck.
Expand 3 Items
Nunc™ TripleFlask™, Cell Culture Flasks, Thermo Scientific
Supplier: Thermo Scientific
The Nunc™ TripleFlask™ is a 3-layer flask providing three times the growth surface of a T175 flask for the same footprint, saving space in the incubator.
Expand 4 Items
Corning® CellBIND® Flasks, Sterile, Corning
Supplier: Corning
CellBIND® surface treatment improves cell attachment by incorporating more oxygen into the cell culture surface, rendering it more hydrophilic and increasing surface stability
Expand 6 Items
Nephelo Culture Flasks with Screw Caps, Ace Glass Incorporated
Supplier: Ace Glass
Nephelo culture flask with side port and angled side arm cuvette for standard nephelometers and spectrophotometers
Expand 7 Items
Flask, Shaker, Ace Glass Incorporated
Supplier: Ace Glass
This shaker flask has a long-style neck for use with plastic closures
Expand 8 Items
Flask, Nephelo Culture, Baffled, Screw Cap, Ace Glass Incorporated
Supplier: Ace Glass
This threaded neck Nephelo culture flask has a clean-out port and a depressed sidearm
Expand 4 Items
Flask, Nephelo Culture, Triple Baffled, Ace Glass Incorporated
Supplier: Ace Glass
This Nephelo culture flask has a long-style neck that accommodates a 25mm stainless steel or plastic closure
Expand 3 Items
Flask, Nephelo Culture, Ace Glass Incorporated
Supplier: Ace Glass
This Nephelo culture flask has a long-style neck that accommodates a 25mm stainless steel or plastic closure
Expand 1 Items
Corning® Elplasia® Microcavity Flask, Corning
Supplier: Corning
Culture thousands of spheroids in a footprint similar to that of a T-75 flask.
Expand 1 Items
Falcon® Cell Culture Flasks, Corning
Supplier: Corning
PS, tissue culture-treated, sterile, non-pyrogenic.
Expand 20 Items
Nunc™ Cell Culture Flasks, Thermo Scientific
Supplier: Thermo Fisher Scientific
These Nunc™ cell culture flasks are designed for culture consistency, cell health, and reproducibility. Choose from a variety of surfaces and sizes with culture areas ranging from 25 to 500 cm² to suit your specific applications and cell types.
Expand 27 Items
Flask, Shaker, Beaded Neck, Ace Glass Incorporated
Supplier: Ace Glass
Shake style culture flask with beaded neck and three bottom edge baffles to enhance mixing and oxygen transfer to media.
Expand 4 Items
Nalgene® Fernbach Culture Flask, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
For growing large-scale cultures
Expand 1 Items
Corning® Polystyrene Tissue Culture Flasks with Vented Caps, Sterile, Corning
Supplier: Corning
These flasks are tissue culture-treated for optimum cell attachment and growth
Expand 5 Items
Trypsinizing Flasks with Pourout and Caps, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
For trypsinizing and decanting cells, the flask is designed for use with magnetic stirrers and spin bars
Expand 5 Items
WHEATON® Nephelo Culture Flasks, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Allows growth and turbidity readings at selected intervals without transfer.
Expand 5 Items
CELLine™ Bioreactor Flasks, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Wheaton manufactures a new line of high density cell culture membrane flasks for antibody and protein production. CELLine works well with secreting cell lines such as Hybridoma, CHO, NS0, SF, HEK, and BHK cells. Conventional in vivo or in vitro cell culture methods can be laborious, low density and require significant purification. CELLine dual compartment flasks separate the cell compartment from the bulk media compartment with a 10 kD MWCO dialysis membrane. This allows for longer run times, high density cultures, and a super concentrated harvest. The CELLine flasks were designed to address three areas of limitation:
Expand 6 Items
Flask, Culture, Screw Cap, Ace Glass Incorporated
Supplier: Ace Glass
This wide mouth flask is designed for culturing organisms requiring a large surface-area-to-volume ratio
Expand 2 Items
Flask, Fernbach, Baffled, Culture, Ace Glass Incorporated
Supplier: Ace Glass
This Fernbach culture flask has a long-style 38mm neck for plastic or stainless steel closures
Expand 2 Items
KIMAX® Culture Flasks, Capacity Scale, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Flasks have a thick-walled, long tubing neck with a plain fire-polished finish sealed to an Erlenmeyer-shaped body
Expand 3 Items
Flask, Shaker, Three Side/Bottom Baffles, Ace Glass Incorporated
Supplier: Ace Glass
These long-style neck flasks have three extra-deep baffles on side and bottom to enhance gas transfer when used with rotary or reciprocating shakers
Expand 5 Items
Falcon® Multi-Flask Cell Culture Flasks, Corning
Supplier: Corning
PS, tissue culture-treated, with membrane filter cap, non-pyrogenic
Expand 2 Items
Trypsinizing Flasks, with Cap, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Wheaton-33® low extractable borosilicate glass composition with narrow lip
Expand 10 Items
KIMBLE® Incubation Flasks, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
These flasks are used for metabolic studies of biological materials. They are particularly useful in measuring C14O2 yields following incubation. Also suitable for in vitro incubations, insulin bio-assays, persulfate oxidations, and the “distillation” of any volatile acid or base.
Expand 2 Items
KIMAX® Baffled Culture Flasks, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Designed for use with rotary and reciprocating shakers where baffle indents provide greater turbulence to improve oxygen transfer rates or aeration