First-Strand cDNA Synthesis Kit, Cytiva
Catalog # CA95040-330L
Supplier: Cytiva
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Description:First-Strand cDNA Synthesis Kit
- Environmentally Preferable:
- Cat. no.:CA95040-330L
- No. of Reactions:55
- Supplier No.:27-9261-01
Specifications
About this item
Designed to generate full length first-strand cDNA from mRNA templates. Following first-strand cDNA synthesis, the sample can be used directly for in vitro amplification using PCR or for second-strand synthesis using the Gubler-Hoffman method.
- For synthesis of single-stranded cDNA prior to amplification by PCR
- Features preassembled bulk reaction mixes to save time and minimise potential for pipetting errors
- Allows users to choose primers
- Includes a choice of two primers: pd(N)6 random hexamers and NotI-(dT)18
Delivery: Kit includes sufficient for up to 55 first-strand syntheses, each of which will produce enough material for up to 15 amplification reactions: Bulk First-Strand cDNA Reaction Mixes, mRNA Standard, DTT Solution, RNase-Free Water, pd(N)6 Primer, NotI-(dT)18 Bifunctional Primer and instruction booklet.
Caution: Storage -15 to -30°C