Order Entry
Canada
ContactUsLinkComponent
 
Amersham ECL Western Blotting Blocking Agent, Cytiva
Catalog # CA95038-568L
Supplier:  Cytiva
CAS Number:  
undefined
Amersham ECL Western Blotting Blocking Agent, Cytiva
Catalog # CA95038-568L
Supplier:  Cytiva
Supplier Number:  RPN2125
CAS Number:  

Specifications

  • Description:
    Amersham ECL Blocking Agent
  • Size:
    40 g
  • Environmentally Preferable:
  • Form:
    Solid
  • Shelf life:
    6 months from manufacture date
  • Storage temperature:
    2 to 8°C
  • Cat. no.:
    CA95038-568L
  • Supplier No.:
    RPN2125
  • No. of Uses:
    20 miniblots

Specifications

About this item

Amersham ECL Blocking Agent is for blocking nitrocellulose and PVDF membranes to be used with Amersham ECL, Amersham ECL Prime and Amersham ECL Select Western Blotting Detection Reagents

Can be used with the Amersham ECL Western blotting detection family. This blocking agent is intended for blocking nitrocellulose and PVDF membranes used with ECL and ECL Plus Western Blotting Detection Reagents.

Delivery: Each pack contains 40 g of membrane blocking agent, sufficient for at least 20 miniblots.