Order Entry
Canada
ContactUsLinkComponent
 
Amersham™ CyDye™ Value Packs (Mono-Reactive Hydrazide), Cytiva
 :  Cytiva
 
undefined
Amersham™ CyDye™ Value Packs (Mono-Reactive Hydrazide), Cytiva
 :  Cytiva
 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Storage conditions:
    2 - 8°C, protected from light; Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles
  • pH insensitive:
    3 pH Insensitive Min./10 pH Insensitive Max. (95056-532 and 95056-502)
  • Shelf life:
    2 weeks (Estimated Shelf Life from Manufacture Date)
  • Solubility:
    Soluble in cold water and hot water
  • Application:
    For labelling via carbonyl groups; For use in steady state fluorescence, FP and FRET applications

 

Frequently Bought Together