PIG® Spill Kit in Small Wall-Mount Cabinet, New Pig
Catalog #: 46000-748
Supplier: New Pig
Specifications
- Cat. no.:46000-748
- Dimensions:Absorbs up to 45.4 L (12 gal.)
- Supplier No.:KIT215
Specifications
About this item
Safety-yellow, heavy-gauge, welded steel cabinet is printed "Spill Kit" for quick identification
- Safety yellow, heavy-gauge, welded steel cabinet is printed "Spill Kit" for quick identification
- Mat and Pillows absorb; Wipers clean surfaces and hands
Prepacked cabinet provides easy access to contents. See-through front panel lets you easily check inventory levels. Includes write-on label for emergency contact info. Two adjustable shelves.
PIG Mat and Pillows absorb; wipers clean surfaces and hands.
Customers Who Bought This Also Bought