PIG® Battery Acid Spill Kit in 20-Gallon Overpack, New Pig
Catalog # 89093-182
Supplier: New Pig
Specifications
- Capacity:Up to 49.2 L (13 gal.)
- Cat. no.:89093-182
- Supplier No.:KIT352
Specifications
About this item
Kit contents are formulated specifically for absorbing and neutralizing battery acid
Includes PPE to guard against corrosive battery acid. 20-gallon overpack salvage drum container meets DOT shipping specifications for overpacking hazardous materials. Temporary disposal bags help make cleanup easier.
No wheels. UN1H2/X75/S, UN1H2/Y100/S.
PIG HazMat Socks stop spreading spills; PIG HazMat Pads and Pillows absorb; Spill-X neutralizes remaining acid.
Frequently Bought Together