ContactUsLinkComponent
PIG® HazMat Spill Kit in 50-Gallon Wheeled Overpack Salvage Drum, New Pig
PIG® HazMat Spill Kit in 50-Gallon Wheeled Overpack Salvage Drum, New Pig
Catalog # 89205-838
Supplier:  New Pig
undefined
PIG® HazMat Spill Kit in 50-Gallon Wheeled Overpack Salvage Drum, New Pig
Catalog # 89205-838
Supplier:  New Pig
Supplier Number:  KIT365
Specifications
  • Capacity:
    Up to 102.2 L (27 gal.)
  • Cat. no.:
    89205-838
  • Supplier No.:
    KIT365
Specifications
About this item
Overpack drum container is UN Rated for shipping waste after spill cleanup
Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail. Lightweight, 100% polyethylene container resists chemicals and keeps contents clean and dry. Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole. Large, color-coded label makes identification fast and accurate. Ledges molded into container make kit easy to move by forklift. Tamperproof seals help prevent pilfering of spill response supplies.
6" polyolefin wheels. UN1H2/X125/S.
6" olefin wheels make spill kit highly mobile for speedy response anywhere in your facility. Lift-out, prepacked baskets speed access and guard contents from UV. PIG HazMat Socks stop spreading spills; PIG HazMat Pads and Pillows absorb quickly. PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide.