PIG® Oil-Only Economy Spill Kits in Duffel Bag, New Pig
Catalog #: 56620-498
Supplier: New Pig
Specifications
- Capacity:Up to 87.1 L (23 gal.)
- Cat. no.:56620-498
- Supplier No.:KIT423
Specifications
About this item
Rugged duffel bag is packed with four kits; absorb spills, then keep the bag
PIG Oil-Only Socks surround and stop spreading spills; PIG Stat-Mat Mat absorbs quickly. Absorbents meet NFPA 99 standards for static decay to allow use with fuels and flammables.
Just open kit, use the absorbents, then throw them in the temporary disposal bags included
Customers Who Bought This Also Bought