PIG® Oil-Only Spill Kit in High-Visibility Bag, New Pig
Catalog # 56620-382
Supplier: New Pig
Specifications
- Capacity:Up to 37.9 L (10 gal.)
- Cat. no.:56620-382
- Supplier No.:KIT420
Specifications
About this item
Durable, water-resistant bag with hook-and-loop closure keeps contents clean and dry
- Temporary disposal bags help make cleanup easier
- Bright-yellow bag and large, color-coded label make the kit easy to locateStrap for easy carrying and hanging
- Compact and easy to store; makes a great truck spill kit
Temporary disposal bags help make cleanup easier. Bright-yellow bag and large, color-coded label make the kit easy to locate. Strap for easy carrying and hanging. Compact and easy to store; makes a great truck spill kit.
PIG Oil-Only Socks stop spreading spills; PIG Stat-Mat Pads absorb quickly. Absorbents meet NFPA 99 standards for static decay to allow use with fuels and flammables
Frequently Bought Together