PIG® Oil-Only Spill Kit in Bucket, New Pig
Catalog #: 89205-844
Supplier: New Pig
Specifications
- Cat. no.:89205-844
- Supplier No.:KIT413
Specifications
About this item
Polyethylene 61/2-gallon bucket features an easy-open, screw-on lid and can be used for shipping or temporarily storing used absorbents
Prepacked bucket provides easy access to contents. Large, color-coded label makes identification fast and accurate. Tamperproof seals help prevent pilfering of spill response supplies. UN Rating 1H2/Y30/S for Packing Groups II and III.
No wheels. UN1H2/Y30/S.
PIG Skimmer Socks stop spreading spills; PIG Stat-Mat Pads absorb quickly. Absorbents meet NFPA 99 standards for static decay to allow use with fuels and flammables
Customers Who Bought This Also Bought