ContactUsLinkComponent
PIG® Spill Kit in Spill Pack, New Pig
PIG® Spill Kit in Spill Pack, New Pig
Catalog # 56620-346
Supplier:  New Pig
undefined
PIG® Spill Kit in Spill Pack, New Pig
Catalog # 56620-346
Supplier:  New Pig
Supplier Number:  KIT261
Specifications
  • Capacity:
    Up to 15.1 L (4 gal.)
  • Cat. no.:
    56620-346
  • Supplier No.:
    KIT261
Specifications
About this item
Rugged vacuum packaging keeps contents clean and dry and makes it easy to detect tampering.
  • Built-in handle and sturdy grommets make the pack easy to carry and hangStores easily behind a vehicle seat
  • Includes nitrile gloves and an extra-large disposal bag with nylon tie to help make cleanup easier
Highly-visible pack is easy to locate. PIG Blue Socks stop spreading spills; PIG Mat absorbs quickly.