- Capacity:16 x 1.5 ml, 8 x 0.5 ml and 8 x 0.2 ml
- Description:Multi-Vortex Mixer
- Electrical:12V
- Tube Ø:16 mm, maximum
- Acceleration:3 sec. (to maximum speed)
- Input power:60 W
- Operating temperature:4...40 °C
- Cat. no.:97025-604
- Orbit:2 mm
- Speed:500 - 3000 rpm
- Supplier No.:V-32
Versatile multi vortex mixer for vigorous re-suspension of cell or chemical pellets in a maximum of 32 tubes (16×1.5 ml,8×0.5 ml and 8×0.2 ml), with the facility to mix individual tubes up to 15 ml
Easy to use; continuous or touch operation and set speed knob from 500 to 3000 rpm.
Orbit: 2 mm. Compact design plus powerful motor gives quiet and consistent performance. Rubber suction pads prevent the unit from ‘walking’ and also absorb vibration.
Supplied with universal 32-place platform and flat platform for single tube mixing. Optional 6 socket platform for 10 ml tubes available separately.
Typical applications include vigorous stirring and re-suspension of bacterial and yeast cells and pellets, metabolite and enzyme extraction and various DNA operations such as deproteinisation of DNA/protein complexes and purification of low molecular weight DNA/RNA fragments.