Order Entry
Canada
ContactUsLinkComponent
PIG® Spill Kit in 30-Gallon Overpack Salvage Drum, New Pig
PIG® Spill Kit in 30-Gallon Overpack Salvage Drum, New Pig
Catalog # 56620-394
Supplier:  New Pig
undefined
PIG® Spill Kit in 30-Gallon Overpack Salvage Drum, New Pig
Catalog # 56620-394
Supplier:  New Pig
Supplier Number:  KIT236

Specifications

  • Capacity:
    Up to 79.5 L (21 gal.)
  • Cat. no.:
    56620-394
  • Supplier No.:
    KIT236

Specifications

About this item

Overpack drum container is UN Rated for shipping waste after spill cleanup

  • Temporary disposal bags help make cleanup easier
  • Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail
  • Lightweight, 100% copolymer container resists chemicals and keeps contents clean and dry
  • Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole
  • Large, color-coded label makes identification fast and accurate
  • Ledges molded into container make kit easy to move by forklift
  • Tamperproof seals help prevent pilfering of spill response supplies

No wheels. UN1H2/X100/S.

Neatly prepacked container speeds access during a spill emergency. PIG Blue Socks stop spreading spills; PIG Mat and Pillows absorb.