Order Entry
Canada
Orders LinkContactUsLinkComponent
 

VWR® Basic Chemically Resistant Aprons

Basic Apron, Yellow, Universal, 57½", 28"×57½", 25½"
  89232-928
VWR® Basic Chemically Resistant Aprons
  89232-928
VWR® Basic Chemically Resistant Aprons
VWR® Basic Chemically Resistant Aprons

Basic Apron, Yellow, Universal, 57½", 28"×57½", 25½"

 
$317.39
 
   
Case of 50 Pallet of 900
 

VWR® Basic chemically resistant aprons provide durable protection against splashes and hazardous chemicals in laboratory and industrial environments. These aprons feature high tensile strength, tear resistance, and compliance with ASTM standards for durability. Designed for comfort and safety, they are latex-free, non-pyrogenic. Ideal for chemical handling, cleaning, and contamination control.

  • Chemical-resistant material for splash and spill protection

  • High tensile strength; meets ASTM D5034 standards for durability
  • Reinforced seams and robust construction for tear resistance and extended use
  • Lightweight, ergonomic fit for long wear without fatigue
  • Latex-free, non-pyrogenic, ISO-certified for cleanroom and lab use
  • Ideal for labs, cleanrooms, chemical handling, and industrial processes
  • Meets regulatory standards while minimizing PPE costs

In environments where chemical splash hazards and contamination risks are part of daily operations, VWR® chemically resistant aprons deliver a critical layer of protection without sacrificing comfort or compliance. These aprons are engineered for laboratories, cleanrooms, and industrial settings where acids, bases, solvents, and other aggressive liquids demand PPE that performs under stress. VWR® aprons are crafted from PVC, PE-coated fabrics, or specialty polymer blends designed to resist penetration by common laboratory chemicals. This multi-layer approach ensures impermeability to liquids, while maintaining flexibility for ease of movement. Reinforced seams and robust edging further enhance durability, making these aprons suitable for repeated use or extended wear in high-risk zones.

Performance is validated through ASTM D5034 tensile strength standards, confirming resilience against tearing and mechanical stress. These garments also meet ISO 13485 and other regulatory benchmarks, ensuring compatibility with controlled environments and GMP protocols. Latex-free and non-pyrogenic properties support sensitive applications, including pharmaceutical and biotech operations.

Despite their rugged chemical resistance, these aprons are designed for operator comfort. Lightweight construction reduces fatigue during long shifts, while adjustable ties and ergonomic patterns allow a secure yet non-restrictive fit. This balance of protection and wearability helps maintain productivity without compromising safety.

 : ISO 13485:2003 Quality Management System. ASTM D5034 Tensile Strength Compliance. FDA Registered Manufacturing Facility. CE Good Distribution Practice. ISO 9001:2015 Certified European Facilities.

 : Aprons are packaged 50 per case.

 
  • Clothing Size:
    Universal
  • Color:
    Yellow
  • Description:
    Basic Apron
  • Length:
    57½"
  • W×L:
    28"×57½"
  • Sleeve:
    25½"
  • Tearing strength:
    816.28 N (machine direction), 769.34 N (cross direction)
  • Seam type:
    Bound
  • Cat. No.:
    89232-928
  • Cleanroom class:
    10000/ISO 7
  • Garment application:
    Cleanroom|Industrial|Laboratory|Food|Chemical protection
  • Gender:
    Unisex
  • Closure type:
    Back fastening
  • Particle shedding (Helmke Drum):
    Level II
  • Water resistance:
    Zero penetration
  • Tensile strength:
    181.20 N (machine direction), 167.30 N (cross direction)
  • Weight:
    88 g/m²