PIG® Skimmer Pillows, New Pig
Supplier: New Pig
Certificates
About this item
Pillow has large surface area, high capacity and fast-wicking filler to quickly soak up oils.
- Absorbs and retains oils and oil-based liquids, including lubricants, fuels and cleaning agents, without taking in a drop of water
- Bright white color makes pillow easier to see in outdoor environments, draws attention to machine leaks and clearly shows saturation level
- Can be incinerated after use to reduce waste or for fuels blending
Tough, chemical-resistant polypropylene skin and filler absorb oils but repel water-based liquids. Polypropylene skin resists chemicals and tearing; reduces dust and holds in liquid, even when saturated. Excellent for skimming oil off of acid baths or for use in other acidic environments. Floats to clean up spills on water confined by booms or socks. Skin is UV resistant up to 12 months; meets NFPA 99 standards for static decay.
Customers Who Bought This Also Bought