Battery Handling PPE Kit, New Pig
89093-184
: New Pig
- For Use With:For PIG® Battery Acid Spill Kit
- Description:Battery Handling PPE Kit
- Environmentally Preferable:
- Cat. no.:89093-184
- Supplier No.:WPL105
Wall-mounted kit holds PPE for handling damaged or leaky batteries.
Includes easy-to-read hydrometer to accurately measure acid concentration. Flashlight requires two AA batteries (not included). All mounting hardware is included.
Includes gloves, goggles, faceshield and apron for protection from battery acid. All components are attached to a high-visibility wall-mountable board labeled "Battery Changing Area.