Tight-Head UN Rated Steel Drum, New Pig
Supplier: New Pig
Certificates
About this item
UN Rated and tested to help ensure safe shipping of hazardous materials. Tight-head design is equipped with 2" NPT and 3/4" NPT bungs.
- Tested to help ensure safe shipping of hazardous materials
- Tight-head design equipped with 2" NPT and 3/4" NPT bungs
Interior is coated with a corrosion inhibitor; choose optional phenolic/epoxy lining for added resistance.18-gauge steel top and bottom with a 20-gauge body meet strict wall thickness requirements to ensure strength, durability and long life.
UN1A1/X1.6/300. 1.2/0.9/1.2 mm
Frequently Bought Together