Nylon Hand Pump, New Pig
Catalog # 56620-596
Supplier: New Pig
Specifications
- Description:For General-Purpose Chemicals
- Cat. no.:56620-596
- Supplier No.:DRM462
Specifications
About this item
Nylon hand pumps fits 15- through 55-gallon drums with 2" NPT bungs.
- Moves 8 oz. of water-based liquid per stroke
- Spout rotates upward to prevent messy drips between uses. Fits
- Maximum recommended viscosity equals SAE 30 weight oil.
Frequently Bought Together