PIG® Traffic Mat® Poly-Back Absorbent Rug, New Pig
Catalog #: 56620-074
Supplier: New Pig
Specifications
- Description:Medium weight
- Dimensions:91.4 cmx30 m (36"x100')
- Supplier No.:MAT223
Specifications
About this item
100% polypropylene material is heat fused and needle punched for incredible durability for walk-on and wheeled traffic
- Absorbs and retains oils, coolants, solvents, and water
- Dark gray color hides grime, so mat stays on the job longer
- Rolls are ideal for walkways and large areas to soak up leaks and overspray
- Cuts easily with scissors or utility knife
- Can be incinerated after use to reduce waste or for fuels blending
- Poly backing prevents absorbed liquids from reaching floors
Our most durable mat won't rip, shred or fray, even under heavy forklift traffic
Customers Who Bought This Also Bought