Order Entry
Canada
ContactUsLinkComponent
PIG® 4-in-1® Absorbent Mat Roll, New Pig
PIG® 4-in-1® Absorbent Mat Roll, New Pig
Catalog # 46000-532
Supplier:  New Pig
undefined
PIG® 4-in-1® Absorbent Mat Roll, New Pig
Catalog # 46000-532
Supplier:  New Pig
Supplier Number:  MAT235

Specifications

  • Description:
    Mediumweight
  • Dimensions:
    40.6 cm x 46 m (16" x 150')
  • Cat. no.:
    46000-532
  • Supplier No.:
    MAT235

Specifications

About this item

Exclusive 4-in-1 Mat is a pad, roll, sock and wiper all in one.

  • Use as a pad to catch drips and soak up spills; roll out to cover large areas; fold along score lines to make a thick, absorbent sock; tear off a piece to use as a low-lint wiper
  • Scuff-resistant, spunbond outer layer resists wear and tear, even under heavy foot traffic or abrasive conditions
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behin
  • Absorbs and retains oils, coolants, solvents and water
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job
  • Rolls are perforated every 10"; easy to tear to size, use only what you need

No matter how you use it, this mat builds on the proven eight-layer, 100% polypropylene construction of PIG Mat.