MacConkey Agar, Hardy Diagnostics
Catalog # CA89407-696
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:Contact plate
- Storage conditions:2-8°C
- Cat. no.:CA89407-696
- Plate Size:15×60 mm
- Volume:16 ml
- Supplier No.:P47
Specifications
About this item
For isolation and differentiation of gram negative bacteria.