Brucella Blood Agar with Hemin, and Vitamin K, Hardy Diagnostics
CA89405-032
:
- Pack type:Plate
- Description:Brucella Blood Agar with Hemin, and Vitamin K
- Size:15×100 mm
- Storage temperature:2 - 8 °C
- Cat. no.:CA89405-032
- Volume:19 ml
- Supplier No.:A30
For the cultivation of anaerobes
A non-inhibitory general growth medium for all gram positive and gram negative anaerobic bacteria. Optimized formula for enhanced growth characteristics of anaerobes.