W-L Nutrient Medium, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
CRITERION™ W-L Nutrient Medium for the cultivation of bacteria and yeasts in brewing and industrial fermentation processes.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Certifications: BSE Free
Specifications
- Storage temperature:–2-30°C