CRITERION™ Tryptose, Hardy Diagnostics
About this item
CRITERION™ Tryptose is primarily used for preparing microbiological culture media and when non-animal source nutrients are needed in the cultivation of fastidious and non-fastidious organsims
- Non-mammalian
- Wide mouth opening
- Reduces dust and allows easy scooping
- Convenient handgrip
- 'No slip' finger holds
- Stackable containers
- High solubility; dissolves quickly
- Custom and bulk products available
Dehydrated culture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
Certifications: TSE/BSE Free
2 Options Available Below
Product Details & Documents
CRITERION™ Tryptose is primarily used for preparing microbiological culture media and when non-animal source nutrients are needed in the cultivation of fastidious and non-fastidious organsims
- Non-mammalian
- Wide mouth opening
- Reduces dust and allows easy scooping
- Convenient handgrip
- 'No slip' finger holds
- Stackable containers
- High solubility; dissolves quickly
- Custom and bulk products available
Dehydrated culture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
Certifications: TSE/BSE Free
Documents
Certificates
Search by Lot NumberHardy Diagnostics Criterion Dehydrated Culture Media
Product Literature
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA89406-666
- CA89406-668
- CA89406-672
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
Specifications
Product Family Options
Product Information
- SizeFinal volumePack typeAvailabilityPrice
- 122.4 g2 LZip-pouch to make 2 L, Mylar®Discontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.CA89406-666Storage temperature–2-30°C - Supplier Number: C8071Specifications:
More
Specifications
Cat. No.CA89406-668Storage temperature–2-30°C - Supplier Number: C8073Specifications:
More
Specifications
Cat. No.CA89406-672Storage temperature–2-30°C