Staphylococcus Select Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Staphylococcus Select Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
CRITERION™ Staphylococcus Selective Agar is a selective medium used for the isolation and differentiation of pathogenic staphylococci
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ Staphylococcus Selective Agar is a selective medium used for the isolation and differentiation of pathogenic staphylococci
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA89406-078
- CA89406-080
- CA89406-082
- CA89406-084
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Specifications
Product Family Options
Product Information
- SizeFinal volumePack typeMinimum Order QuantityAvailabilityPrice
- Supplier Number: C7000Specifications:
More
Specifications
Cat. No.CA89406-078Storage temperature–2-30°C - Supplier Number: C7001Specifications:
More
Specifications
Cat. No.CA89406-080Storage temperature–2-30°C - Supplier Number: C7002Specifications:
More
Specifications
Cat. No.CA89406-082Storage temperature–2-30°C - Supplier Number: C7003Specifications:
More
Specifications
Cat. No.CA89406-084Storage temperature–2-30°C