Phenol Red Dexrose Broth, CRITERION™, Hardy Diagnostics
Catalog # CA89406-650
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:Zip-pouch to make 2 L, Mylar®
- Size:122.4 g
- Storage temperature:–2-30°C
- Cat. no.:CA89406-650
- Final Volume:2 L
- Supplier No.:C8050
Specifications
About this item
CRITERION™ Phenol Red Dextrose Broth for the differentiation of microorganisms by dextrose fermentation
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free