MacConkey Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
CRITERION™ MacConkey Broth is recommended for cultivating gram-negative, lactose-fermenting bacilli from water and foods as a presumptive test for coliforms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
MacConkey Broth can also be used preenrichment of E. coli O157 for toxin testing. Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
- Storage temperature:–2-30°C
Specifications
Related Information
Certificates
Search by Lot Number Frequently Bought Together