Letheen Broth, Modified, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
CRITERION™ Letheen Broth, Modified, for the determination of antimicrobial activity of quaternary ammonium compounds
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
- Storage temperature:2-8°C