EMB Agar, H, H & T, CRITERION™, Hardy Diagnostics
CRITERION™ EMB Agar, HHT is recommended for the isolation and identification of gram-negative enteric bacteria
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
- Storage temperature:–2-30°C
Frequently Bought Together