Orders LinkContactUsLinkComponent
EC Medium, CRITERION™, Hardy Diagnostics
EC Medium, CRITERION™, Hardy Diagnostics
EC Medium, CRITERION™, Hardy Diagnostics
Certificates
EC Medium, CRITERION™, Hardy Diagnostics
2 Options Available Below
Product Details & Documents
CRITERION™ EC Medium is recommended for the detection of coliforms at 35 degrees C
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
and Escherichia coli and fecal coliforms at 45.5 degrees C. Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • CA89405-422
  • CA89405-424
  • Storage temperature:
  • –2-30°C
  • –2-30°C
Specifications
Product Family Options
Product Information
  • Size
    Pack type
    Availability
    Price
  • 500 g
    Bottle, wide mouth
    Log in for availability
    Log in for availability
    Each
    Each - $381.00
    $381.00
    Each
       $381.00
    Minimum order quantity is 1
    Supplier Number: C5691
    Specifications:
    More
    Specifications
    Cat. No.
    CA89405-422
    Storage temperature
    –2-30°C
  • 2 kg
    Bucket
    Log in for availability
    Log in for availability
    Each
    Each - $1,480.00
    $1,480.00
    Each
       $1,480.00
    Minimum order quantity is 1
    Supplier Number: C5692
    Specifications:
    More
    Specifications
    Cat. No.
    CA89405-424
    Storage temperature
    –2-30°C