Pseudomonas Isolation Agar, 15×100 mm plate, Hardy Diagnostics
Catalog # CA89407-226
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:Plate
- Description:Isolation Agar
- Storage conditions:2-8°C
- Minimum Order Quantity:10
- Cat. no.:CA89407-226
- Plate Size:15×100 mm
- Volume:18 ml
- Supplier No.:G219