Order Entry
Canada
Orders LinkContactUsLinkComponent
GN Broth, Hardy Diagnostics

GN Broth, Hardy Diagnostics

GN Broth, 16 x 125 mm, 10 ml, Tube
Catalog # CA89407-602
Supplier #:  K39
GN Broth, Hardy Diagnostics
Catalog # CA89407-602
CAS Number:  
GN Broth, Hardy Diagnostics
GN Broth, Hardy Diagnostics

GN Broth, 16 x 125 mm, 10 ml, Tube

CAS Number:  
Price
$95.60
Log in for availability
Pack of 20
Minimum order quantity is 1
Product Details & Documents

For the selective enrichment of Salmonella and Shigella species in stool specimens.

Documents

Specifications

Specifications

  • Pack type:
    Tube
  • Description:
    GN Broth
  • Size:
    16 x 125 mm
  • Fill volume:
    10 ml
  • Cat. No.:
    CA89407-602
  • Storage temperature:
    –2-30°C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.