Order Entry
Canada
Orders LinkContactUsLinkComponent
Fluid D, USP

Fluid D, USP

Fluid D, USP
Certificates
Fluid D, USP

2 Options Available Below

Product Details & Documents

Fluid D is a rinsing and diluting fluid for use in USP testing protocols.

  • Rinsing and diluting fluid
  • Contains peptic digest of animal tissue and polysorbate 80

A rinsing and diluting fluid for use in USP testing protocols. pH 7.1. Contains peptic digest of animal tissue and polysorbate 80.

Product Family Options

Product Information

  • Size
    Pack type
    Storage temperature
    Availability
    Price
  • 100 ml
    Glass bottle, septum-sealed
    –2...30 °C
    Log in for availability
    Log in for availability
    Pack of 20
    Pack of 20 - $329.00
    $329.00
    Pack of 20
       $329.00
    Minimum order quantity is 1

    Supplier Number: U110
  • 1 L
    PC bottle, narrow mouth, screw top
    –2...30 °C
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $467.00
    $467.00
    Pack of 10
       $467.00

    Supplier Number: U115
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.