Dichloran Rose Bengal Chloramphenicol Agar
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
DRBC for use in the cultivation of yeasts and molds associated with food, dietary supplements and other matrices.
- Optically clear polystyrene - For distortion free viewing
- Heavy-Duty - Reduces breakage during shipping
- Stacking rings - Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
- Breathable Packaging
Packaging: Packaged in sleeves of 10 plates in a 'breathable' bag that prevents buildup of condensation and excess moisture.
Caution: This product is not intended to be used for the diagnosis of human disease.
Specifications
- Storage temperature:2...8 °C