DG-18 Agar (Dichloran-Glycerol), Hardy Diagnostics
Catalog # CA89407-488
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:Plate
- Size:15 x 100 mm
- Storage temperature:2-8°C
- Cat. no.:CA89407-488
- Volume:26 ml
- Supplier No.:W85
Specifications
About this item
For the selective isolation and cultivation of xerophilic molds.