Order Entry
Canada
Orders LinkContactUsLinkComponent
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate

Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate

Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
 
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate

 

 

Recommended for use as general purpose growth media for the isolation, cultivation, and differentiation of a wide variety of microorganisms.

  • Optimized for superior performance
  • For the non-selective cultivation of microorganisms
  • General growth medium for bacteria, yeast, and fungi
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
  • Packaged in sleeves of 10 plates, in 'breathable' cellulose bags that prevents build-up of condensation and excess moisture
  • The 100 plate size box offers superior protection during shipment and is easy to open without a cutting tool

Contains 5% sheep blood from a veterinary supervised, controlled herd of donor animals. Blood is guaranteed to be from healthy antibiotic-free animals.

 

 

 

  • Catalog No:
  • CA76047-180
  • CA89405-024
  • Storage conditions:
  • 2...8 °C
  • 2...8 °C

 

Product Family Options

 

  • Volume
    Application
    Plate size
    Size
     
     
  • 19 ml
    Clinical
    1.5×10 cm
    100/case, 10/pack
       
       
    Pack of 100
    Pack of 100 - $355.00
    $355.00
    Pack of 100
       $355.00
    Minimum order quantity is 1

     : A10BX
     
     
     
    Specifications
    Cat. No.
    CA76047-180
    Storage conditions
    2...8 °C
  • 19 ml
    Clinical
    1.5×10 cm
    10/pack
       
       
    Pack of 10
    Pack of 10 - $38.90
    $38.90
    Pack of 10
       $38.90
    Minimum order quantity is 1

     : A10
     
     
     
    Specifications
    Cat. No.
    CA89405-024
    Storage conditions
    2...8 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.