ContactUsLinkComponent
Bacillus Cereus Selective Agar, Hardy Diagnostics
  CA89407-178
 :  
undefined
Bacillus Cereus Selective Agar, Hardy Diagnostics
  CA89407-178
 :  G156
 :  
  • Pack type:
    Plate
  • Size:
    15 x 100 mm
  • Environmentally Preferable:
  • Storage temperature:
    2-8°C
  • Minimum Order Quantity:
    10
  • Cat. no.:
    CA89407-178
  • Supplier No.:
    G156
A selective medium which detects small numbers of Bacillus cereus cells and spores in the presence of large numbers of other contaminating organisms.