Tryptose Broth, CRITERION™, Hardy Diagnostics
CRITERION™ Tryptose Broth is used for cultivating a wide variety of fastidious microorganisms, particularly Brucella spp.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
: BSE Free
- Storage temperature:–2-30°C
Frequently Bought Together