Orders LinkContactUsLinkComponent
Thioglycollate Medium without Indicator, CRITERION™, Hardy Diagnostics
Thioglycollate Medium without Indicator, CRITERION™, Hardy Diagnostics
Thioglycollate Medium without Indicator, CRITERION™, Hardy Diagnostics
Certificates
Thioglycollate Medium without Indicator, CRITERION™, Hardy Diagnostics
Thioglycollate Medium without Indicator, CRITERION™, Hardy Diagnostics
4 Options Available Below
Product Details & Documents
CRITERION™ Thioglycollate Medium without Indicator is recommended for the cultivation of aerobic, microaerophilic, and anaerobic microorganisms
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • CA89406-134
  • CA89406-136
  • CA89406-138
  • CA89406-140
  • Storage temperature:
  • –2-30°C
  • –2-30°C
  • –2-30°C
  • –2-30°C
Specifications
Product Family Options
Product Information
  • Size
    Final volume
    Pack type
    Availability
    Price
  • 122.4 g
    2 L
    Zip-pouch to make 2 L, Mylar®
    Log in for availability
    Log in for availability
    Each
    Each - $122.00
    $122.00
    Each
       $122.00
    Minimum order quantity is 1
    Supplier Number: C7080
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-134
    Storage temperature
    –2-30°C
  • 500 g
    Bottle, wide mouth
    Log in for availability
    Log in for availability
    Each
    Each - $181.00
    $181.00
    Each
       $181.00
    Minimum order quantity is 1
    Supplier Number: C7081
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-136
    Storage temperature
    –2-30°C
  • 2 kg
    Bucket
    Log in for availability
    Log in for availability
    Each
    Each - $633.00
    $633.00
    Each
       $633.00
    Minimum order quantity is 1
    Supplier Number: C7082
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-138
    Storage temperature
    –2-30°C
  • 10 kg
    Bucket
    Log in for availability
    Log in for availability
    Each
    Each - $2,400.00
    $2,400.00
    Each
       $2,400.00
    Minimum order quantity is 1
    Supplier Number: C7083
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-140
    Storage temperature
    –2-30°C