Order Entry
Canada
ContactUsLinkComponent
Rappaport-Vassiliadis R10 Broth, Hardy Diagnostics
Rappaport-Vassiliadis R10 Broth, Hardy Diagnostics
Catalog # CA89405-956
CAS Number:  
undefined
Rappaport-Vassiliadis R10 Broth, Hardy Diagnostics
Catalog # CA89405-956
Supplier Number:  C6731
CAS Number:  

Specifications

  • Pack type:
    Bottle, wide mouth
  • Size:
    500 g
  • Storage temperature:
    –2-30°C
  • Cat. no.:
    CA89405-956
  • Supplier No.:
    C6731

Specifications

About this item

CRITERION™ Rappaport-Vassiliadis R10 Broth is primarily for use in the selective enrichment and isolation of Salmonella spp

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications

from food and environmental specimens.Dehydratedculture media with engineered container.

Certifications: BSE Free