Order Entry
Canada
Orders LinkContactUsLinkComponent
 

CRITERION™ Phosphate Buffer, pH 7.2

CRITERION™ Phosphate Buffer, pH 7.2
Certificates
CRITERION™ Phosphate Buffer, pH 7.2

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CRITERION™ Agar, bacteriological grade is used as a solidifying agent in the preparation of solid and semi-solid culture media.

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • Increased solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
  • BSE-free
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • CA89406-750
  • CA76196-446
  • Storage temperature:
  • 2...30 °C
  • 2...30 °C

Specifications

Product Family Options

Product Information

  • Size
    Pack type
    Application
    Availability
    Price
  • 500 g
    Bottle, wide mouth
    Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing
    Log in for availability
    Log in for availability
    Each
    Each - $178.00
    $178.00
    Each
       $178.00
     

    Supplier Number: C8331
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-750
    Storage temperature
    2...30 °C
  • 10 kg
    Bucket
    Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing
    Log in for availability
    Log in for availability
    Each (10kg)
    Each (10kg) - $2,050.00
    $2,050.00
    Each (10kg)
       $2,050.00
     

    Supplier Number: C8333
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    CA76196-446
    Storage temperature
    2...30 °C