Peptic Digest Agar Base, CRITERION™, Hardy Diagnostics
CRITERION™ Peptic Digest Agar Base, an enrichment medium recommended for the selective isolation of Haemophilus spp
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
CRITERION™ Peptic Digest Agar Base, an enrichment medium recommended for the selective isolation of Haemophilus spp
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
- Catalog No:
- CA89405-858
- CA89405-860
- CA89405-862
- CA89405-864
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Product Family Options
- SizeFinal volumePack type
- : C6560
Specifications
Cat. No.CA89405-858Storage temperature–2-30°C - : C6561
Specifications
Cat. No.CA89405-860Storage temperature–2-30°C - : C6562
Specifications
Cat. No.CA89405-862Storage temperature–2-30°C - : C6563
Specifications
Cat. No.CA89405-864Storage temperature–2-30°C