CRITERION™ Papaic Digest of Soy Bean Meal, Hardy Diagnostics
CRITERION™ Papaic Digest of Soybean Meal is an enzymatic soybean meal prepared especially for use in microbiological cultures.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
It is recommended for use in media for the cultivation of a large variety of organisms, including fungi, and is also used in media for microbiological assay. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
: BSE Free
- Storage temperature:–2-30°C
Frequently Bought Together