Nutrient Broth, CRITERION™, Hardy Diagnostics
CRITERION™ Nutrient Broth is a general purpose growth media recommended for use in the cultivation of nonfastidious microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
: BSE Free
- Storage temperature:–2-30°C
Frequently Bought Together