Orders LinkContactUsLinkComponent
Mycoplasma Broth Base, CRITERION™, Hardy Diagnostics
Mycoplasma Broth Base, CRITERION™, Hardy Diagnostics
Mycoplasma Broth Base, CRITERION™, Hardy Diagnostics
Certificates
Mycoplasma Broth Base, CRITERION™, Hardy Diagnostics
Mycoplasma Broth Base, CRITERION™, Hardy Diagnostics
5 Options Available Below
Product Details & Documents
CRITERION™ Mycoplasma Broth Base is for the isolation and cultivation of Mycoplasma spp
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Mycoplasma Supplement, product #283610, is required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • CA89406-616
  • CA89406-618
  • CA89406-620
  • CA89406-622
  • CA89406-624
  • Storage temperature:
  • –2-30°C
  • –2-30°C
  • –2-30°C
  • –2-30°C
  • –2-30°C
Specifications
Product Family Options
Product Information
  • Size
    Final volume
    Pack type
    Availability
    Price
  • 122.4 mL
    2 L
    Zip-pouch to make 2 L, Mylar®
    Log in for availability
    Log in for availability
    Each
    Each - $117.00
    $117.00
    Each
       $117.00
    Minimum order quantity is 1
    Supplier Number: C8000
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-616
    Storage temperature
    –2-30°C
  • 500 g
    Bottle, wide mouth
    Log in for availability
    Log in for availability
    Each
    Each - $239.00
    $239.00
    Each
       $239.00
    Minimum order quantity is 1
    Supplier Number: C8001
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-618
    Storage temperature
    –2-30°C
  • 2 kg
    Bucket
    Log in for availability
    Log in for availability
    Each
    Each - $855.00
    $855.00
    Each
       $855.00
    Minimum order quantity is 1
    Supplier Number: C8002
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-620
    Storage temperature
    –2-30°C
  • 10 kg
    Bucket
    Log in for availability
    Log in for availability
    Each
    Each - $3,340.00
    $3,340.00
    Each
       $3,340.00
    Minimum order quantity is 1
    Supplier Number: C8003
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-622
    Storage temperature
    –2-30°C
  • 50 kg
    Drum
    Log in for availability
    Log in for availability
    Each
    Each - $15,450.00
    $15,450.00
    Each
       $15,450.00
    Minimum order quantity is 1
    Supplier Number: C8004
    Specifications:
    More
    Specifications
    Cat. No.
    CA89406-624
    Storage temperature
    –2-30°C