Modified Plate Count Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
CRITERION™ Modified Plate Count Agar, (Plate Count Agar with Bromcresol Purple and Tween® 80), is recommended for the cultivation and differentiation of Lactobacilli
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
- Storage conditions:–2-30°C
Specifications
Related Information
Certificates
Search by Lot Number Frequently Bought Together